Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(1R,3S)-3-Hydroxy-1-(hydroxymethyl)-3-phenylpropyl]dodecanamide 98.0+%, TCI America™
SDP

Catalog No. H15535MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15535MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H15535MG Supplier TCI America Supplier No. H15535MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 383418-30-2
Molecular Formula C22H37NO3
Molecular Weight (g/mol) 363.54
MDL Number MFCD29472535
InChI Key YCAKBKAOFSILDC-UHFFFAOYNA-N
Synonym HPA-12, N-[(2R,4S)-1,4-Dihydroxy-4-phenylbutan-2-yl]dodecanamide
PubChem CID 11079004
IUPAC Name N-(1,4-dihydroxy-4-phenylbutan-2-yl)dodecanamide
SMILES CCCCCCCCCCCC(=O)NC(CO)CC(O)C1=CC=CC=C1

Specifications

Color White
Quantity 5 mg
Formula Weight 363.54
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-[(1R,3S)-3-Hydroxy-1-(hydroxymethyl)-3-phenylpropyl]dodecanamide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.