Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-[(1R,8S,9S)-Bicyclo[6.1.0]Non-4-Yn-9-Ylmethyloxycarbonyl]-1,8-Diamino-3,6-Dioxaoctane

Catalog No. 745073100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-507-3100MG 100 mg
74-507-325MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 74-507-3100MG Supplier Sigma Aldrich Supplier No. 745073100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-[(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyloxycarbonyl]-1,8-diamino-3,6-dioxaoctane may be used to functionalize hyaluronic acid with bicyclo[6.1.0]nonyne (BCN) for the development of responsive HA-based physical hydrogels with tunable properties.

Specifications

CAS 1263166-93-3
Molecular Formula C17H28N2O4
Linear Formula C17H28N2O4
MDL Number MFCD19705418
Quantity 100 mg
Synonym (1alpha,8alpha,9beta)-Bicyclo[6.1.0]non-4-yn-9-ylmethyl N-{2-[2-(2-aminoethoxy)ethoxy]ethyl}carbamate; BCN-amine
Molecular Weight (g/mol) 324.42 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.