Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[(1S)-1-[3-(4-ethoxyphenyl)-3,4-dihydro-4-oxopyrido[2,3-d]pyrimidin-2-yl]ethyl]-N-(3-pyridinylmethyl | 473720-30-8 | 99.9% | 603.59 g/mol | C32H28F3N5O4 | 10 MG
SDP

Catalog No. 5000286210
Encompass_Preferred

AMG 487 (S-enantiomer) is the S enantiomer of AMG 487, a small-molecule antagonist of the CXC chemokine receptor 3 (CXCR3) supplied for research use. The compound is provided as a solid and is characterized by a defined chemical formula, molecular weight, CAS registry number, high purity, and recommended storage conditions.

  • S enantiomer of a selective CXCR3 antagonist.
  • High purity (99.94%).
  • Chemical formula C32H28F3N5O4.
  • Molecular weight 603.59 g/mol.
  • CAS number 473720-30-8.
  • Off-white to light yellow solid appearance.
  • Recommended storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (2 years) or -20 °C (1 year).

Catalog No. 50-002-86210 Supplier Medchemexpress LLC Supplier No. HY15319B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AMG 487 (S-enantiomer) is the S enantiomer of AMG 487, a small-molecule antagonist of the CXC chemokine receptor 3 (CXCR3) supplied for research use. The compound is provided as a solid and is characterized by a defined chemical formula, molecular weight, CAS registry number, high purity, and recommended storage conditions.

  • S enantiomer of a selective CXCR3 antagonist.
  • High purity (99.94%).
  • Chemical formula C32H28F3N5O4.
  • Molecular weight 603.59 g/mol.
  • CAS number 473720-30-8.
  • Off-white to light yellow solid appearance.
  • Recommended storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (2 years) or -20 °C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.