Learn More
Medchemexpress LLC N-[(1S,2R)-1-(3-fluoro-2-methylphenyl)-1-hydroxypentan-2-yl]-2-oxo-1,3-dihydroindole-4-carboxamide | 2406205-61-4 | 99.8% | C21H23FN2O3 | 10MG
SDP
Catalog No. 5000260083
Encompass_Preferred
A1AT modulator 1 is a small-molecule inhibitor of Z α1-antitrypsin polymerization with reported potency (pIC50 = 8.3). The compound is supplied as a white to off-white solid for research use, has molecular formula C21H23FN2O3, molecular weight 370.42 g/mol, and an analytical purity of approximately 99.8%.
  • Exhibits potent inhibition of Z α1-antitrypsin polymerization (pIC50 = 8.3).
  • High analytical purity suitable for in vitro research.
  • Supplied as a solid in small milligram pack sizes for assay flexibility.
  • Storage recommendations provided for powder and solution to preserve stability.
  • Applicable to biochemical and cell-based studies of serpin modulation.
Catalog No. 50-002-60083 Supplier Medchemexpress LLC Supplier No. HY13485810MG
A1AT modulator 1 is a small-molecule inhibitor of Z α1-antitrypsin polymerization with reported potency (pIC50 = 8.3). The compound is supplied as a white to off-white solid for research use, has molecular formula C21H23FN2O3, molecular weight 370.42 g/mol, and an analytical purity of approximately 99.8%.
  • Exhibits potent inhibition of Z α1-antitrypsin polymerization (pIC50 = 8.3).
  • High analytical purity suitable for in vitro research.
  • Supplied as a solid in small milligram pack sizes for assay flexibility.
  • Storage recommendations provided for powder and solution to preserve stability.
  • Applicable to biochemical and cell-based studies of serpin modulation.