Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[(1S,2R,3E)-1-(hydroxymethyl)-2-hydroxy-3-heptadecen-1-yl]tetracosanamide | 34435-05-7 | 99.9% | 650.11 g/mol | C42H83NO3 | 5 MG
SDP

Catalog No. 5000223383
Encompass_Preferred

C24-ceramide is a long-chain ceramide (N-lignoceroyl-D-erythro-sphingosine) used in biochemical research to investigate sphingolipid signaling, apoptosis, and mTOR pathway modulation. Supplied as a white to off-white solid, it is intended for in vitro and cellular studies and is soluble in ethanol and THF with ultrasonic assistance.

  • Purity of 99.9% for reliable experimental consistency.
  • White to off-white solid form for easy handling and storage.
  • Soluble in ethanol and THF with sonication for preparation of stock solutions.
  • Stable when stored frozen as powder to preserve integrity.
  • Suitable for studies of PIP4K2C, mTOR signaling, and sphingolipid biology.

Catalog No. 50-002-23383 Supplier Medchemexpress LLC Supplier No. HY1345085MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C24-ceramide is a long-chain ceramide (N-lignoceroyl-D-erythro-sphingosine) used in biochemical research to investigate sphingolipid signaling, apoptosis, and mTOR pathway modulation. Supplied as a white to off-white solid, it is intended for in vitro and cellular studies and is soluble in ethanol and THF with ultrasonic assistance.

  • Purity of 99.9% for reliable experimental consistency.
  • White to off-white solid form for easy handling and storage.
  • Soluble in ethanol and THF with sonication for preparation of stock solutions.
  • Stable when stored frozen as powder to preserve integrity.
  • Suitable for studies of PIP4K2C, mTOR signaling, and sphingolipid biology.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.