Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-((1S,2S)-1-(3,5-Di-Tert-Butylphenyl)-2-(Quinolin-2-Yl)Butyl)Acetamide

Catalog No. ALD00596 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ALD00596 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ALD00596 Supplier Sigma Aldrich Supplier No. ALD00596100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This chiral acetyl-protected aminoethyl quinoline (APAQ) ligand was developed by the Yu group for ligand-enabled acceleration of Pd-catalyzed enantioselective beta-methylene C-H functionalization, which is important for the generation of beta-chiral centers in asymmetric synthesis.Ligand-accelerated enantioselective methylene C(sp3)-H bond activation

Specifications

CAS 555-06-6
Molecular Formula C24H20N2O6
Linear Formula C24H20N2O6
MDL Number MFCD01317717
Quantity 1 g
Molecular Weight (g/mol) 432.43 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.