Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[(1S)-5-(acetylamino)-1-[[(4-methyl-2-oxo-2H-1-benzopyran-7-yl)amino]carbonyl]pentyl]-carbamic acid | 233691-67-3 | MFCD02683946 | 99.5% | 445.51 g/mol | C23H31N3O6 | 5 MG
SDP

Catalog No. 5000222902
Encompass_Preferred

Boc-Lys(Ac)-AMC is a cell-permeable fluorogenic substrate used to measure histone deacetylase (HDAC) activity in enzymatic and cell-based assays. After HDAC-mediated deacetylation and protease cleavage, the AMC fluorophore is released and quantified by fluorescence (Ex/Em = 355 nm/460 nm), enabling sensitive HDAC activity detection and inhibitor screening.

  • Cell-permeable fluorometric HDAC substrate.
  • Fluorescence readout at Ex/Em = 355 nm/460 nm.
  • High purity suitable for biochemical assays.
  • Available as a solid or as a 10 mM solution in DMSO.
  • High DMSO solubility (≈100 mg/mL) with sonication.
  • Storage recommended at -20°C; in solvent store at -80°C for long term.
  • Supplied in small research quantities, including 5 mg packs.

Catalog No. 50-002-22902 Supplier Medchemexpress LLC Supplier No. HY1381595MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Lys(Ac)-AMC is a cell-permeable fluorogenic substrate used to measure histone deacetylase (HDAC) activity in enzymatic and cell-based assays. After HDAC-mediated deacetylation and protease cleavage, the AMC fluorophore is released and quantified by fluorescence (Ex/Em = 355 nm/460 nm), enabling sensitive HDAC activity detection and inhibitor screening.

  • Cell-permeable fluorometric HDAC substrate.
  • Fluorescence readout at Ex/Em = 355 nm/460 nm.
  • High purity suitable for biochemical assays.
  • Available as a solid or as a 10 mM solution in DMSO.
  • High DMSO solubility (≈100 mg/mL) with sonication.
  • Storage recommended at -20°C; in solvent store at -80°C for long term.
  • Supplied in small research quantities, including 5 mg packs.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.