Learn More
Medchemexpress LLC N-(2-(1H-indol-3-yl)ethyl)-5-(5-fluoropyridin-3-yl)-3-methylpyrazolo[1,5-a]pyrimidin-7-amine | 2247950-73-6 | 98.0% | 386.42 g/mol | C22H19FN6 | 5 MG
SDP
Catalog No. 5000223459
Encompass_Preferred
KYN-101 is a potent, selective, orally active aryl hydrocarbon receptor (AHR) inhibitor used in research. It reduces CYP1A1 mRNA expression and has demonstrated anti-cancer activity in preclinical studies. Supplied as a small-quantity research compound with accompanying quality documentation.
  • Potent, selective aryl hydrocarbon receptor (AHR) inhibitor.
  • Orally active in preclinical models.
  • Decreases CYP1A1 mRNA expression in cellular assays.
  • Demonstrates anti-cancer activity in preclinical studies.
  • Supplied as a small research quantity (e.g., 5 mg) with COA and datasheet.
  • High purity (approximately 98.0%).
  • For research use only; not for human or clinical use.
Catalog No. 50-002-23459 Supplier Medchemexpress LLC Supplier No. HY1342175MG
KYN-101 is a potent, selective, orally active aryl hydrocarbon receptor (AHR) inhibitor used in research. It reduces CYP1A1 mRNA expression and has demonstrated anti-cancer activity in preclinical studies. Supplied as a small-quantity research compound with accompanying quality documentation.
  • Potent, selective aryl hydrocarbon receptor (AHR) inhibitor.
  • Orally active in preclinical models.
  • Decreases CYP1A1 mRNA expression in cellular assays.
  • Demonstrates anti-cancer activity in preclinical studies.
  • Supplied as a small research quantity (e.g., 5 mg) with COA and datasheet.
  • High purity (approximately 98.0%).
  • For research use only; not for human or clinical use.