Learn More
Medchemexpress LLC N-(2-(1H-indol-3-yl)ethyl)-5-(5-fluoropyridin-3-yl)-3-methylpyrazolo[1,5-a]pyrimidin-7-amine | 2247950-73-6 | 98.0% | C22H19FN6 | 10MG
SDP
Catalog No. 5000261386
Encompass_Preferred
KYN-101 is a small-molecule, selective, orally active antagonist of the aryl hydrocarbon receptor (AHR) supplied for laboratory research. It reduces CYP1A1 mRNA expression and has demonstrated anti-cancer activity in preclinical studies, making it suitable for mechanistic studies of AHR signaling and oncology research.
  • Selective AHR inhibition with reported nanomolar potency.
  • Orally active in preclinical models.
  • Reduces CYP1A1 mRNA expression in cellular assays.
  • Useful for mechanistic studies of AHR signaling and cancer biology.
  • Available as solid and ready-to-use DMSO solutions for assay setup.
  • High purity suitable for research applications.
Catalog No. 50-002-61386 Supplier Medchemexpress LLC Supplier No. HY13421710MG
KYN-101 is a small-molecule, selective, orally active antagonist of the aryl hydrocarbon receptor (AHR) supplied for laboratory research. It reduces CYP1A1 mRNA expression and has demonstrated anti-cancer activity in preclinical studies, making it suitable for mechanistic studies of AHR signaling and oncology research.
  • Selective AHR inhibition with reported nanomolar potency.
  • Orally active in preclinical models.
  • Reduces CYP1A1 mRNA expression in cellular assays.
  • Useful for mechanistic studies of AHR signaling and cancer biology.
  • Available as solid and ready-to-use DMSO solutions for assay setup.
  • High purity suitable for research applications.