Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)ethyl)-12-(4-(2-fluoro-5-((4-oxo... | 2523016-96-6 | 99.9% | 876.97 g/mol | C47H53FN8O8 | 5 MG
SDP

Catalog No. 5000223326
Encompass_Preferred

SK-575 is a proteolysis-targeting chimera (PROTAC) that selectively induces degradation of PARP1 and is used in biochemical and cellular research to study PARP1 biology and cancer cell responses.

  • Potent PARP1 degrader with reported IC50 2.30 nM.
  • High purity (99.86%).
  • Molecular weight 876.97 g/mol.
  • Molecular formula C47H53FN8O8.
  • Soluble in DMSO at ~80 mg/mL; ultrasonic assistance recommended.
  • Supplied in small research quantities (for example, 5 mg).

Catalog No. 50-002-23326 Supplier Medchemexpress LLC Supplier No. HY1391565MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SK-575 is a proteolysis-targeting chimera (PROTAC) that selectively induces degradation of PARP1 and is used in biochemical and cellular research to study PARP1 biology and cancer cell responses.

  • Potent PARP1 degrader with reported IC50 2.30 nM.
  • High purity (99.86%).
  • Molecular weight 876.97 g/mol.
  • Molecular formula C47H53FN8O8.
  • Soluble in DMSO at ~80 mg/mL; ultrasonic assistance recommended.
  • Supplied in small research quantities (for example, 5 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.