Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide 98.0+%, TCI America™
SDP

Catalog No. T27135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T27135G Supplier TCI America Supplier No. T27135G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 66065-85-8
Molecular Formula C7H6Cl3NO5
Molecular Weight (g/mol) 290.477
MDL Number MFCD00075216
InChI Key WBZXNGAFYBGQFE-UHFFFAOYSA-N
Synonym 2,5-dioxopyrrolidin-1-yl 2,2,2-trichloroethyl carbonate, succinimidyl 2,2,2-trichloroethyl carbonate, n-2,2,2-trichloroethoxycarbonyloxy succinimide, troc-osu, succinimidyl-2,2,2-trichloroethyl carbonate, succinimidyl 2,2,2-trichloroethyl carbon, zlchem 876, acmc-20aajg, n-succinimidyl 2,2,2-trichloroethyl carbonate
PubChem CID 4192178
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 2,2,2-trichloroethyl carbonate
SMILES C1CC(=O)N(C1=O)OC(=O)OCC(Cl)(Cl)Cl

Specifications

Melting Point 113°C
Color White
Quantity 5 g
Formula Weight 290.48
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.