Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N'-[[2-(2,3-dichlorophenoxy)phenyl]methyl]-N,N'-dimethylethane-1,2-diamine | 2289726-31-2 | >98.0% | 375.72 g/mol | C17H21Cl3N2O | 10 MG
SDP

Catalog No. 5000288707
Encompass_Preferred

DCPT1061 hydrochloride is a research-use small-molecule inhibitor of protein arginine methyltransferases, reported to inhibit PRMT1, PRMT6, and PRMT8. Supplied as the hydrochloride salt to improve handling and solubility, it is provided in small milligram quantities for laboratory use. Typical distributor-reported specifications include a molecular weight of 375.72 g/mol, formula C17H21Cl3N2O, and reported purity greater than 98%. Store sealed at -20°C and prepare solutions in DMSO as required.

  • Reported inhibitor of PRMT1, PRMT6, and PRMT8.
  • Supplied as the hydrochloride salt for improved solubility and handling.
  • Molecular weight 375.72 g/mol and formula C17H21Cl3N2O.
  • Purity reported greater than 98% by distributors.
  • Soluble in DMSO (~50 mg/mL) with warming or sonication.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-88707 Supplier Medchemexpress LLC Supplier No. HY162003A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DCPT1061 hydrochloride is a research-use small-molecule inhibitor of protein arginine methyltransferases, reported to inhibit PRMT1, PRMT6, and PRMT8. Supplied as the hydrochloride salt to improve handling and solubility, it is provided in small milligram quantities for laboratory use. Typical distributor-reported specifications include a molecular weight of 375.72 g/mol, formula C17H21Cl3N2O, and reported purity greater than 98%. Store sealed at -20°C and prepare solutions in DMSO as required.

  • Reported inhibitor of PRMT1, PRMT6, and PRMT8.
  • Supplied as the hydrochloride salt for improved solubility and handling.
  • Molecular weight 375.72 g/mol and formula C17H21Cl3N2O.
  • Purity reported greater than 98% by distributors.
  • Soluble in DMSO (~50 mg/mL) with warming or sonication.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.