Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-5-yl)-2-(trifluoromethoxy)benzenesulfonamide | 2765625-93-0 | 98.4% | C20H14F3N3O7S | 10MG
SDP

Catalog No. 5000260601
Encompass_Preferred

SJ6986 is a potent, selective, and orally active molecular glue degrader of GSPT1 and GSPT2 that induces robust target degradation (DC50 = 2.1 nM; Dmax = 99%). It is supplied for research use and characterized for in vitro and in vivo formulation and stability.

  • Potent GSPT1/2 degradation with DC50 = 2.1 nM and Dmax = 99%.
  • Orally bioavailable small molecule suitable for in vivo studies.
  • High purity: 98.4%.
  • Molecular formula C20H14F3N3O7S, molecular weight 497.40 g/mol.
  • Soluble in DMSO (100 mg/mL) and ≥2.5 mg/mL in 10% DMSO/90% SBE-β-CD saline formulation.
  • Stable as powder at -20°C (3 years) and 4°C (2 years); in solution store at -80°C (6 months).

Catalog No. 50-002-60601 Supplier Medchemexpress LLC Supplier No. HY13219910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SJ6986 is a potent, selective, and orally active molecular glue degrader of GSPT1 and GSPT2 that induces robust target degradation (DC50 = 2.1 nM; Dmax = 99%). It is supplied for research use and characterized for in vitro and in vivo formulation and stability.

  • Potent GSPT1/2 degradation with DC50 = 2.1 nM and Dmax = 99%.
  • Orally bioavailable small molecule suitable for in vivo studies.
  • High purity: 98.4%.
  • Molecular formula C20H14F3N3O7S, molecular weight 497.40 g/mol.
  • Soluble in DMSO (100 mg/mL) and ≥2.5 mg/mL in 10% DMSO/90% SBE-β-CD saline formulation.
  • Stable as powder at -20°C (3 years) and 4°C (2 years); in solution store at -80°C (6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.