Learn More
Medchemexpress LLC N-(2-(2-acrylamidophenyl)-4-(pyridin-4-yl)thiazol-5-yl)-2-(difluoromethyl)benzamide | 2227368-54-7 | MFCD34593626 | 99.0% | 452.46 g/mol | C25H20N6O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY103017A25MG
Encompass_Preferred
Potent and selective IRAK1 inhibitor for research use. The compound inhibits IRAK1 with an apparent IC50 of 9 nM and shows no inhibition of IRAK4 at concentrations up to 10 μM. Supplied as a solid or as a DMSO solution, it provides defined solubility and storage properties.
  • Potent, selective IRAK1 inhibition with an apparent IC50 of 9 nM.
  • High purity (99.03%) for reliable experimental results.
  • Available as solid and as 10 mM solutions in DMSO for flexible dosing.
  • Molecular weight 452.46 g/mol; molecular formula C25H20N6O3.
  • High solubility in DMSO (125 mg/mL) and in vivo formulation solubility ≥1.98 mg/mL under protocol conditions.
  • Stable storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-47368