Learn More
Medchemexpress LLC N-(2-((2-amino-4-thiazolyl)amino)ethyl)-N'-isopropyl-S-methylisothiourea. | 210354-22-6 | 98.4% | 100 MG

Supplier: Medchemexpress LLC HY12119100MG
GW274150 is a selective inducible nitric oxide synthase (iNOS) inhibitor used in biochemical and cellular research. It is supplied as a high-purity small-molecule solid with documented molecular properties and recommended storage conditions.
- Selective iNOS inhibitor used in biochemical and cellular research.
- High purity: 98.4%.
- Molecular formula C8H17N3O2S; molecular weight 219.30.
- Supplied as a 100 mg solid for laboratory use.
- Recommended storage: 4°C; in solvent: -80°C for 6 months or -20°C for 1 month (store under nitrogen).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.