Learn More
Medchemexpress LLC N-[2-(2-benzyl-1H-indol-3-yl)ethyl]acetamide | 117946-91-5 | MFCD00672498 | 100.0% | 292.37 g/mol | C19H20N2O | 10 MG
SDP
Catalog No. 5000213226
Encompass_Preferred
Luzindole is a selective melatonin receptor antagonist used in research to probe melatonin signaling and receptor function. It preferentially binds the MT2 receptor over MT1 (reported Ki values ~10.2 nM for MT2 and ~158 nM for MT1) and has been used in preclinical studies affecting circadian, mood, and immune responses. Supplied as an off-white to light-yellow solid with reported high purity, it is typically offered in small research quantities.
  • Selective melatonin receptor antagonist useful for receptor pharmacology studies.
  • Higher affinity for MT2 versus MT1 with reported Ki values.
  • Reported molecular weight 292.37 g/mol and formula C19H20N2O.
  • Supplied as an off-white to light-yellow solid with high reported purity.
  • Suitable for preclinical and in vitro research applications.
Catalog No. 50-002-13226 Supplier Medchemexpress LLC Supplier No. HY10125410MG
Luzindole is a selective melatonin receptor antagonist used in research to probe melatonin signaling and receptor function. It preferentially binds the MT2 receptor over MT1 (reported Ki values ~10.2 nM for MT2 and ~158 nM for MT1) and has been used in preclinical studies affecting circadian, mood, and immune responses. Supplied as an off-white to light-yellow solid with reported high purity, it is typically offered in small research quantities.
  • Selective melatonin receptor antagonist useful for receptor pharmacology studies.
  • Higher affinity for MT2 versus MT1 with reported Ki values.
  • Reported molecular weight 292.37 g/mol and formula C19H20N2O.
  • Supplied as an off-white to light-yellow solid with high reported purity.
  • Suitable for preclinical and in vitro research applications.