Learn More
Medchemexpress LLC N-[2-(2-iodo-5-methoxy-1H-indol-3-yl)ethyl]acetamide | 93515-00-5 | MFCD00055216 | 99.0% | 358.17 | C13H15IN2O2 | 200 MG
SDP
Catalog No. 5000211247
Encompass_Preferred
2-Iodomelatonin is a halogenated melatonin analogue supplied as a research reagent and analytical standard. It acts as a high-affinity agonist at melatonin receptor 1 (MT1) with marked selectivity over MT2, and is used to identify, characterize, and localize melatonin binding sites in neural and peripheral tissues.
Catalog No. 50-002-11247 Supplier Medchemexpress LLC Supplier No. HY101176200MG
2-Iodomelatonin is a halogenated melatonin analogue supplied as a research reagent and analytical standard. It acts as a high-affinity agonist at melatonin receptor 1 (MT1) with marked selectivity over MT2, and is used to identify, characterize, and localize melatonin binding sites in neural and peripheral tissues.