Learn More
Medchemexpress LLC N-[2-[(3,4-dichlorophenyl)amino]-4-quinolinyl]cyclohexanecarboxamide | 1116652-18-6 | MFCD18251557 | 99.2% | 414.33 | 1 MG
SDP
Catalog No. 5000344774
Encompass_Preferred
LUF6096 is a potent allosteric enhancer of the adenosine A3 receptor, which is able to allosterically enhance agonist binding. It shows low orthosteric affinity for any of the adenosine receptors. LUF6096 also demonstrates protective effects in myocardial ischemia/reperfusion injury.
  • Enhances agonist binding to adenosine A3 receptor
  • Exhibits low orthosteric affinity for adenosine receptors
  • Provides protective effects in myocardial ischemia/reperfusion injury
Catalog No. 50-003-44774 Supplier Medchemexpress LLC Supplier No. HY1091550MG
LUF6096 is a potent allosteric enhancer of the adenosine A3 receptor, which is able to allosterically enhance agonist binding. It shows low orthosteric affinity for any of the adenosine receptors. LUF6096 also demonstrates protective effects in myocardial ischemia/reperfusion injury.
  • Enhances agonist binding to adenosine A3 receptor
  • Exhibits low orthosteric affinity for adenosine receptors
  • Provides protective effects in myocardial ischemia/reperfusion injury