Learn More
Medchemexpress LLC N-(2,3-dihydro-1,4-benzodioxin-3-ylmethyl)-2-(2,6-dimethoxyphenoxy)ethanamine; hydrochloride | 2170-58-3 | MFCD00055187 | 97.1% | 381.85 | C19H24ClNO5 | 5 MG
SDP
Catalog No. 5000264608
Encompass_Preferred
(±)-WB 4101 hydrochloride is the racemic hydrochloride salt of WB 4101, a selective α-adrenergic (noradrenaline) receptor antagonist used in research to block norepinephrine-mediated responses. It is supplied as a solid of high purity for pharmacological and biochemical studies of adrenergic receptor function.
  • Selective α-adrenergic receptor antagonist.
  • Inhibits norepinephrine-mediated contraction in tissue assays.
  • Supplied as a solid with high purity for reliable results.
  • Available in small pack sizes suitable for preparative studies.
  • Applicable to receptor pharmacology and smooth muscle research.
Catalog No. 50-002-64608 Supplier Medchemexpress LLC Supplier No. HY12718A5MG
(±)-WB 4101 hydrochloride is the racemic hydrochloride salt of WB 4101, a selective α-adrenergic (noradrenaline) receptor antagonist used in research to block norepinephrine-mediated responses. It is supplied as a solid of high purity for pharmacological and biochemical studies of adrenergic receptor function.
  • Selective α-adrenergic receptor antagonist.
  • Inhibits norepinephrine-mediated contraction in tissue assays.
  • Supplied as a solid with high purity for reliable results.
  • Available in small pack sizes suitable for preparative studies.
  • Applicable to receptor pharmacology and smooth muscle research.