Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(2-((4-(1,1-difluoroethyl)oxazol-2-yl)methyl)-2H-1,2,3-triazol-4-yl)-2-methyl-5-(m-tolyl)oxazole-4 | 1258417-54-7 | 100.0% | 428.39 | C20H18F2N6O3 | 100MG
SDP

Catalog No. 5000235773
Encompass_Preferred

ACT-389949 is a potent, selective agonist of formyl peptide receptor 2 (FPR2/ALX) used in pharmacological research on inflammatory pathways. It shows an EC50 of 3 nM for FPR2/ALX internalization and is supplied as a high-purity solid with recommended solvent and formulation guidance for in vitro and in vivo studies.

  • Potent FPR2/ALX agonist with an EC50 of 3 nM.
  • High reported purity (~99.96%).
  • Soluble in DMSO; in vivo formulation protocols available.
  • Available in multiple pack sizes from 1 mg to 500 mg.
  • Molecular formula C20H18F2N6O3; molecular weight 428.39.

Catalog No. 50-002-35773 Supplier Medchemexpress LLC Supplier No. HY124071100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACT-389949 is a potent, selective agonist of formyl peptide receptor 2 (FPR2/ALX) used in pharmacological research on inflammatory pathways. It shows an EC50 of 3 nM for FPR2/ALX internalization and is supplied as a high-purity solid with recommended solvent and formulation guidance for in vitro and in vivo studies.

  • Potent FPR2/ALX agonist with an EC50 of 3 nM.
  • High reported purity (~99.96%).
  • Soluble in DMSO; in vivo formulation protocols available.
  • Available in multiple pack sizes from 1 mg to 500 mg.
  • Molecular formula C20H18F2N6O3; molecular weight 428.39.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.