Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[2-[4-(1,3-benzodioxol-5-yl)-2-(1,1-dimethylethyl)phenoxy]ethyl]-N-methylglycine hydrochloride | 1779796-27-8 | ≥98.0% | 421.91 g/mol | C22H28ClNO5 | 10 MG
SDP

Catalog No. 5000213907
Encompass_Preferred

LY2365109 hydrochloride is a selective small-molecule inhibitor of the glycine transporter 1 (GlyT1). It blocks glycine uptake in hGlyT1-expressing cells with an IC50 of 15.8 nM and is supplied as the hydrochloride salt for research use in neurochemical and pharmacological studies.

  • Selective GlyT1 inhibition (IC50 15.8 nM)
  • Hydrochloride salt, solid form
  • High purity ≥98% (HPLC)
  • Molecular weight 421.91 g/mol
  • CAS 1779796-27-8
  • Suitable for neurochemical and pharmacological research applications

Catalog No. 50-002-13907 Supplier Medchemexpress LLC Supplier No. HY100416A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LY2365109 hydrochloride is a selective small-molecule inhibitor of the glycine transporter 1 (GlyT1). It blocks glycine uptake in hGlyT1-expressing cells with an IC50 of 15.8 nM and is supplied as the hydrochloride salt for research use in neurochemical and pharmacological studies.

  • Selective GlyT1 inhibition (IC50 15.8 nM)
  • Hydrochloride salt, solid form
  • High purity ≥98% (HPLC)
  • Molecular weight 421.91 g/mol
  • CAS 1779796-27-8
  • Suitable for neurochemical and pharmacological research applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.