Learn More
Medchemexpress LLC N-(2-(4-(3,5-Dioxo-1,2,4-triazolidin-4-yl)-2-methoxyphenoxy)ethyl)-4,7,10,13-tetraoxahexadec-ynamide | 99.3% | 508.52 g/mol | C23H32N4O9 | 50 MG
SDP
Catalog No. 5000204168
Encompass_Preferred
PTAD-PEG4-alkyne is a cleavable four-unit polyethylene glycol linker containing a PTAD moiety and a terminal alkyne. It is intended for bioconjugation and antibody-drug conjugate synthesis, enabling tyrosine-selective conjugation and copper-catalyzed azide-alkyne cycloaddition.
  • Cleavable PEG4 linker enabling controlled release
  • PTAD moiety for tyrosine-selective conjugation
  • Terminal alkyne compatible with CuAAC click chemistry
  • High purity (99.3%) suitable for research use
  • Soluble in DMSO (125 mg/mL); store powder at -20°C
Catalog No. 50-002-04168 Supplier Medchemexpress LLC Supplier No. HY13604650MG
PTAD-PEG4-alkyne is a cleavable four-unit polyethylene glycol linker containing a PTAD moiety and a terminal alkyne. It is intended for bioconjugation and antibody-drug conjugate synthesis, enabling tyrosine-selective conjugation and copper-catalyzed azide-alkyne cycloaddition.
  • Cleavable PEG4 linker enabling controlled release
  • PTAD moiety for tyrosine-selective conjugation
  • Terminal alkyne compatible with CuAAC click chemistry
  • High purity (99.3%) suitable for research use
  • Soluble in DMSO (125 mg/mL); store powder at -20°C