Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2,4,6-Trichlorophenyl)maleimide 98.0+%, TCI America™
SDP

Catalog No. T198725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T198725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T198725G Supplier TCI America Supplier No. T198725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13167-25-4
Molecular Formula C10H4Cl3NO2
Molecular Weight (g/mol) 276.497
MDL Number MFCD06797173
InChI Key VHZJMAJCUAWIHV-UHFFFAOYSA-N
PubChem CID 83195
IUPAC Name 1-(2,4,6-trichlorophenyl)pyrrole-2,5-dione
SMILES C1=CC(=O)N(C1=O)C2=C(C=C(C=C2Cl)Cl)Cl

Specifications

Melting Point 134°C
Color White
Quantity 25 g
Formula Weight 276.50
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(2,4,6-Trichlorophenyl)maleimide
EINECSNumber (5)-5426
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.