Learn More
Medchemexpress LLC N-(2,4-difluorophenyl)-2-(4-(2-(4-ethylsulfonylphenyl)acetamido)-2-fluorophenyl)-2-methylpropanamide | 2068119-11-7 | 99.3% | 50 MG
SDP
Catalog No. 5000232556
Encompass_Preferred
S18-000003 is a potent, selective, orally active inhibitor of RORγt intended for preclinical research. It demonstrates sub-30 nM activity against human RORγt in binding assays and shows strong selectivity versus other ROR family members. The compound is supplied as a solid with high purity and is soluble in DMSO for biochemical and cellular studies.
  • Potent RORγt inhibition (IC50 <30 nM).
  • High selectivity over other ROR family members (IC50 >10 μM).
  • High purity of 99.31% suitable for research applications.
  • Soluble in DMSO for solution preparation.
  • Available in mg-scale quantities for assay development.
Catalog No. 50-002-32556 Supplier Medchemexpress LLC Supplier No. HY11936650MG
S18-000003 is a potent, selective, orally active inhibitor of RORγt intended for preclinical research. It demonstrates sub-30 nM activity against human RORγt in binding assays and shows strong selectivity versus other ROR family members. The compound is supplied as a solid with high purity and is soluble in DMSO for biochemical and cellular studies.
  • Potent RORγt inhibition (IC50 <30 nM).
  • High selectivity over other ROR family members (IC50 >10 μM).
  • High purity of 99.31% suitable for research applications.
  • Soluble in DMSO for solution preparation.
  • Available in mg-scale quantities for assay development.