Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(2,4-difluorophenyl)-2-[4-[[2-(4-EtSO2-phenyl)acetyl]amino]-2-fluorophenyl]-2-methylpropanamide | 2068119-11-7 | 99.3% | 25 MG
SDP

Catalog No. 5000233781
Encompass_Preferred

A potent, selective, orally active RORγt inhibitor intended for research use, exhibiting high affinity for human RORγt and suitability for cell and animal studies.

  • Potent RORγt inhibition (IC50 <30 nM).
  • High selectivity over other ROR family members (IC50 >10 μM).
  • High purity: 99.31%.
  • Molecular weight 518.55; formula C26H25F3N2O4S.
  • Available in multiple sizes including 25 mg; also supplied as a 10 mM solution in DMSO.
  • Optimized solubility and vehicle options for cell and animal experiments.
  • Storage: powder -20°C (3 years); in solvent -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-33781 Supplier Medchemexpress LLC Supplier No. HY11936625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A potent, selective, orally active RORγt inhibitor intended for research use, exhibiting high affinity for human RORγt and suitability for cell and animal studies.

  • Potent RORγt inhibition (IC50 <30 nM).
  • High selectivity over other ROR family members (IC50 >10 μM).
  • High purity: 99.31%.
  • Molecular weight 518.55; formula C26H25F3N2O4S.
  • Available in multiple sizes including 25 mg; also supplied as a 10 mM solution in DMSO.
  • Optimized solubility and vehicle options for cell and animal experiments.
  • Storage: powder -20°C (3 years); in solvent -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.