Learn More
Medchemexpress LLC N-(2,4-dimethoxyphenyl)-N-(1-oxo-2-propyn-1-yl)-2-(2-thienyl)glycyl-glycine ethyl ester | 1401089-31-3 | 99.3% | 430.47 | C21H22N2O6S | 50 MG
SDP
Catalog No. 5000214161
Encompass_Preferred
PACMA 31 is an irreversible small-molecule inhibitor of protein disulfide isomerase (PDI) used for biochemical and cell-based research. It covalently modifies PDI active-site cysteines, has a reported IC50 of about 10 μM, shows tumor-targeting activity in preclinical models, and contains an alkyne group compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAC) click chemistry.
  • Irreversible PDI inhibitor that covalently modifies active-site cysteines.
  • Reported IC50 of approximately 10 μM for PDI activity inhibition.
  • Demonstrated tumor-targeting activity in preclinical studies.
  • Contains an alkyne functional group for CuAAC click chemistry applications.
  • Solid form and high reported purity suitable for research use.
Catalog No. 50-002-14161 Supplier Medchemexpress LLC Supplier No. HY10043350MG
PACMA 31 is an irreversible small-molecule inhibitor of protein disulfide isomerase (PDI) used for biochemical and cell-based research. It covalently modifies PDI active-site cysteines, has a reported IC50 of about 10 μM, shows tumor-targeting activity in preclinical models, and contains an alkyne group compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAC) click chemistry.
  • Irreversible PDI inhibitor that covalently modifies active-site cysteines.
  • Reported IC50 of approximately 10 μM for PDI activity inhibition.
  • Demonstrated tumor-targeting activity in preclinical studies.
  • Contains an alkyne functional group for CuAAC click chemistry applications.
  • Solid form and high reported purity suitable for research use.