Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(2,4-dimethoxyphenyl)-N-(1-oxo-2-propyn-1-yl)-2-(2-thienyl)glycyl-glycine, ethyl ester | 1401089-31-3 | 99.3% | 430.47 g/mol | C21H22N2O6S | 25 MG
SDP

Catalog No. 5000214896
Encompass_Preferred

PACMA 31 is an irreversible, orally active small-molecule inhibitor of protein disulfide isomerase (PDI) used as a research reagent in biochemical and cellular studies. It covalently modifies active-site cysteines and has shown antitumor activity in preclinical models. For research use only; not for human or clinical applications.

  • Irreversible inhibitor of protein disulfide isomerase
  • Forms covalent bond with active-site cysteines
  • High purity (99.3%) suitable for assays
  • Molecular weight 430.47 g/mol; formula C21H22N2O6S
  • Available in 25 MG vial for laboratory use
  • CAS number 1401089-31-3 for substance identification

Catalog No. 50-002-14896 Supplier Medchemexpress LLC Supplier No. HY10043325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PACMA 31 is an irreversible, orally active small-molecule inhibitor of protein disulfide isomerase (PDI) used as a research reagent in biochemical and cellular studies. It covalently modifies active-site cysteines and has shown antitumor activity in preclinical models. For research use only; not for human or clinical applications.

  • Irreversible inhibitor of protein disulfide isomerase
  • Forms covalent bond with active-site cysteines
  • High purity (99.3%) suitable for assays
  • Molecular weight 430.47 g/mol; formula C21H22N2O6S
  • Available in 25 MG vial for laboratory use
  • CAS number 1401089-31-3 for substance identification

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.