Learn More
Medchemexpress LLC N-(2,4-dimethylphenyl)-2,2,2-trifluoro-N-[(E)-(3-methoxyphenyl)methylideneamino]acetamide | 1146963-51-0 | MFCD25976644 | 99.8% | C18H17F3N2O2 | 10MG
SDP
Catalog No. 5000259460
Encompass_Preferred
(E/Z)-J147 (N-(2,4-dimethylphenyl)-2,2,2-trifluoro-N-[(E)-(3-methoxyphenyl)methylideneamino]acetamide) is a small-molecule research compound investigated for neuroprotective and cognitive-enhancing effects. It readily crosses the blood-brain barrier and has reported activity against monoamine oxidase B and the dopamine transporter. The material is supplied as a high-purity solid for laboratory use.
  • High purity: 99.8% by supplier specification.
  • Powder form, off-white to pink in color.
  • Recommended solvent: DMSO soluble at ≥100 mg/mL.
  • Storage: powder stable at -20°C (long term) and 4°C (short term).
  • Demonstrated neuroprotective activity and blood-brain barrier penetration.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-59460 Supplier Medchemexpress LLC Supplier No. HY1377910MG
(E/Z)-J147 (N-(2,4-dimethylphenyl)-2,2,2-trifluoro-N-[(E)-(3-methoxyphenyl)methylideneamino]acetamide) is a small-molecule research compound investigated for neuroprotective and cognitive-enhancing effects. It readily crosses the blood-brain barrier and has reported activity against monoamine oxidase B and the dopamine transporter. The material is supplied as a high-purity solid for laboratory use.
  • High purity: 99.8% by supplier specification.
  • Powder form, off-white to pink in color.
  • Recommended solvent: DMSO soluble at ≥100 mg/mL.
  • Storage: powder stable at -20°C (long term) and 4°C (short term).
  • Demonstrated neuroprotective activity and blood-brain barrier penetration.
  • For research use only; not for human or clinical use.