Learn More
Medchemexpress LLC N-(2,4-dinitrophenyl)glycine | 1084-76-0 | MFCD00024400 | 99.6% | 241.16 g/mol | C8H7N3O6 | 250 MG
SDP
Catalog No. 5000228336
Encompass_Preferred
N-(2,4-dinitrophenyl)glycine is a high-purity amino acid derivative supplied as a 250 mg solid for laboratory research. It is commonly used as a derivatized glycine reagent in biochemical and pharmacological studies. The compound has molecular formula C8H7N3O6, molecular weight 241.16 g/mol, and CAS number 1084-76-0. Store protected from light at 4°C; in solution, store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • High purity: 99.6% as stated by supplier.
  • Available as a 250 MG solid pack suitable for bench-scale experiments.
  • Well characterized: molecular weight 241.16 g/mol; formula C8H7N3O6.
  • Suitable as a derivatized amino acid reagent in biochemical assays.
  • Light-sensitive: protect from light during storage and handling.
Catalog No. 50-002-28336 Supplier Medchemexpress LLC Supplier No. HY118174250MG
N-(2,4-dinitrophenyl)glycine is a high-purity amino acid derivative supplied as a 250 mg solid for laboratory research. It is commonly used as a derivatized glycine reagent in biochemical and pharmacological studies. The compound has molecular formula C8H7N3O6, molecular weight 241.16 g/mol, and CAS number 1084-76-0. Store protected from light at 4°C; in solution, store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • High purity: 99.6% as stated by supplier.
  • Available as a 250 MG solid pack suitable for bench-scale experiments.
  • Well characterized: molecular weight 241.16 g/mol; formula C8H7N3O6.
  • Suitable as a derivatized amino acid reagent in biochemical assays.
  • Light-sensitive: protect from light during storage and handling.