Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(2-((5-chloro-2-((2-methoxy-4-(4-methylpiperazin-1-yl)phenyl)amino)pyrimidin-4-yl)amino)phenyl)... | 2254805-62-2 | 98.6% | 518.03 g·mol⁻¹ | C23H28ClN7O3S | 5 MG
SDP

Catalog No. 5000222562
Encompass_Preferred

ZX-29 is a research-grade small-molecule ALK inhibitor described as an autophagy inducer and apoptosis promoter in cellular studies. It is supplied for laboratory research use only and is not for human, veterinary, or therapeutic use.

  • Research-grade small-molecule ALK inhibitor and autophagy inducer.
  • Induces apoptosis by endoplasmic reticulum stress and can overcome ALK mutation-driven resistance.
  • High purity reported (approximately 98.6%).
  • Molecular weight 518.03 g·mol⁻¹ and formula C23H28ClN7O3S.
  • Soluble in DMSO; supplied in multiple small-scale pack sizes including 5 mg.

Catalog No. 50-002-22562 Supplier Medchemexpress LLC Supplier No. HY1358875MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZX-29 is a research-grade small-molecule ALK inhibitor described as an autophagy inducer and apoptosis promoter in cellular studies. It is supplied for laboratory research use only and is not for human, veterinary, or therapeutic use.

  • Research-grade small-molecule ALK inhibitor and autophagy inducer.
  • Induces apoptosis by endoplasmic reticulum stress and can overcome ALK mutation-driven resistance.
  • High purity reported (approximately 98.6%).
  • Molecular weight 518.03 g·mol⁻¹ and formula C23H28ClN7O3S.
  • Soluble in DMSO; supplied in multiple small-scale pack sizes including 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.