Learn More
Medchemexpress LLC N-[2,6-bis(1-methylethyl)phenyl]-N'-[4-[(4-nitrophenyl)thio]phenyl]urea | 228544-65-8 | 99.7% | 449.57 g/mol | C25H27N3O3S | 50 MG
SDP
Catalog No. 5000240039
Encompass_Preferred
VULM 1457 is a small-molecule research compound described as a potent inhibitor of cholesterol acyltransferase (acyl-CoA). It reduces production and secretion of adrenomedullin, down-regulates adrenomedullin receptors in human hepatoblastic cells, and exhibits hypolipidaemic activity and protective effects in myocardial ischemia-reperfusion models. Intended for research use only.
  • Potent inhibitor of cholesterol acyltransferase (acyl-CoA).
  • Reduces production and secretion of adrenomedullin.
  • Down-regulates adrenomedullin receptors in hepatoblastic cells.
  • Demonstrates hypolipidaemic activity.
  • Shows protective effects in myocardial ischemia-reperfusion models.
  • Provided as a high-purity research reagent for laboratory use.
Catalog No. 50-002-40039 Supplier Medchemexpress LLC Supplier No. HY10757150MG
VULM 1457 is a small-molecule research compound described as a potent inhibitor of cholesterol acyltransferase (acyl-CoA). It reduces production and secretion of adrenomedullin, down-regulates adrenomedullin receptors in human hepatoblastic cells, and exhibits hypolipidaemic activity and protective effects in myocardial ischemia-reperfusion models. Intended for research use only.
  • Potent inhibitor of cholesterol acyltransferase (acyl-CoA).
  • Reduces production and secretion of adrenomedullin.
  • Down-regulates adrenomedullin receptors in hepatoblastic cells.
  • Demonstrates hypolipidaemic activity.
  • Shows protective effects in myocardial ischemia-reperfusion models.
  • Provided as a high-purity research reagent for laboratory use.