Learn More
Medchemexpress LLC N-[2,6-bis(1-methylethyl)phenyl]-N'-[4-[(4-nitrophenyl)thio]phenyl]urea | 228544-65-8 | MFCD10687104 | 99.7% | 449.57 g/mol | C25H27N3O3S | 5 MG
SDP
Catalog No. 5000240171
Encompass_Preferred
VULM 1457 is a research-grade small-molecule inhibitor of acyl-CoA (cholesterol acyltransferase) intended for biochemical and cellular studies. It is supplied as a solid at high reported purity (99.7%), with molecular weight 449.57 g/mol and formula C25H27N3O3S, and is provided in small pack sizes for assay development.
  • High purity suitable for analytical and biological assays.
  • Supplied in small pack sizes for assay development.
  • Compatible with DMSO stock solution preparation for cellular testing.
  • Useful for studying cholesterol esterification and related pathways.
  • Documentation available for handling and preparation.
Catalog No. 50-002-40171 Supplier Medchemexpress LLC Supplier No. HY1075715MG
VULM 1457 is a research-grade small-molecule inhibitor of acyl-CoA (cholesterol acyltransferase) intended for biochemical and cellular studies. It is supplied as a solid at high reported purity (99.7%), with molecular weight 449.57 g/mol and formula C25H27N3O3S, and is provided in small pack sizes for assay development.
  • High purity suitable for analytical and biological assays.
  • Supplied in small pack sizes for assay development.
  • Compatible with DMSO stock solution preparation for cellular testing.
  • Useful for studying cholesterol esterification and related pathways.
  • Documentation available for handling and preparation.