Learn More
Medchemexpress LLC N-(2,6-dichlorophenyl)-N'-(5-nitro-3-thienyl)urea | 672286-03-2 | 99.5% | 332.16 g/mol | C11H7Cl2N3O3S | 10MG
SDP
Catalog No. 5000238250
Encompass_Preferred
DFP00173 is a potent, selective small-molecule inhibitor of aquaporin-3 (AQP3) intended for research use. It inhibits mouse and human AQP3 with an IC50 of approximately 0.1-0.4 μM and reduces erythrocyte glycerol permeability (IC50 ≈ 0.2 μM). The compound is supplied as a high-purity solid with documented solubility and storage guidelines for in vitro and in vivo studies.
  • Potent AQP3 inhibition at submicromolar concentrations.
  • Selective for AQP3 over related AQP isoforms.
  • High purity suitable for biochemical and cellular assays.
  • Readily soluble in DMSO for in vitro dosing.
  • Validated in vivo formulation protocols available.
  • Stable under recommended powder storage conditions.
Catalog No. 50-002-38250 Supplier Medchemexpress LLC Supplier No. HY12607310MG
DFP00173 is a potent, selective small-molecule inhibitor of aquaporin-3 (AQP3) intended for research use. It inhibits mouse and human AQP3 with an IC50 of approximately 0.1-0.4 μM and reduces erythrocyte glycerol permeability (IC50 ≈ 0.2 μM). The compound is supplied as a high-purity solid with documented solubility and storage guidelines for in vitro and in vivo studies.
  • Potent AQP3 inhibition at submicromolar concentrations.
  • Selective for AQP3 over related AQP isoforms.
  • High purity suitable for biochemical and cellular assays.
  • Readily soluble in DMSO for in vitro dosing.
  • Validated in vivo formulation protocols available.
  • Stable under recommended powder storage conditions.