Learn More
Medchemexpress LLC N-(2,6-dimethylphenyl)-2-(4-(2-hydroxy-3-(2-methoxyphenoxy)propyl)piperazin-1-yl-2,2,3,3,5,5,6,6-d8)acetamide dihydrochloride | 1219802-60-4 | 99.0% | 508.51 | 1 MG
SDP
Catalog No. 5000385755
Encompass_Preferred
Ranolazine-d8 (dihydrochloride) is the deuterium labeled Ranolazine dihydrochloride. Ranolazine dihydrochloride (CVT 303 dihydrochloride) is an anti-angina agent that achieves its effects by inhibiting the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively) without affecting heart rate or blood pressure (BP). Ranolazine dihydrochloride is also a partial fatty acid oxidation inhibitor. It is for research use only and not sold to patients.
  • Isotope-labeled compounds
  • Hot research area
  • Internal standard for clinical mass spectrometry
  • Isotope-labeled inhibitors
  • Deuterium (2H, D) stable isotope-labeled inhibitors
  • Stable isotope-labeled inhibitors for use in in vivo cell experiments
  • Therapeutic drug monitoring: TDM
Catalog No. 50-003-85755 Supplier Medchemexpress LLC Supplier No. HY17401S1MG
Ranolazine-d8 (dihydrochloride) is the deuterium labeled Ranolazine dihydrochloride. Ranolazine dihydrochloride (CVT 303 dihydrochloride) is an anti-angina agent that achieves its effects by inhibiting the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively) without affecting heart rate or blood pressure (BP). Ranolazine dihydrochloride is also a partial fatty acid oxidation inhibitor. It is for research use only and not sold to patients.
  • Isotope-labeled compounds
  • Hot research area
  • Internal standard for clinical mass spectrometry
  • Isotope-labeled inhibitors
  • Deuterium (2H, D) stable isotope-labeled inhibitors
  • Stable isotope-labeled inhibitors for use in in vivo cell experiments
  • Therapeutic drug monitoring: TDM