Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2,6-Dimethylphenyl)thiourea, 99%, Thermo Scientific™

Catalog No. AAL1299806
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1299806 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1299806 Supplier Thermo Scientific Chemicals Supplier No. L1299806

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6396-76-5
Odor Odorless
Molecular Formula C9H12N2S
Quantity 5g
MDL Number MFCD00041165
UN Number UN2811
Beilstein 2209836
Synonym 1-2,6-dimethylphenyl thiourea, 1-2,6-dimethylphenyl-2-thiourea, 2,6-dimethylphenyl thiourea, n-2,6-dimethylphenyl thiourea, 1-2,6-xylyl thiourea, thiourea, 2,6-dimethylphenyl, amino 2,6-dimethylphenyl amino methane-1-thione, acmc-20apd5, 1-2,6-xylyl-2-thiourea, 2,6-dimethyl-phenyl-thiourea
InChI Key ASNKJUONFPQYPC-UHFFFAOYSA-N
SMILES CC1=CC=CC(C)=C1NC(N)=S
IUPAC Name (2,6-dimethylphenyl)thiourea
Molecular Weight (g/mol) 180.27
PubChem CID 853911
Formula Weight 180.27
Percent Purity 99%
Chemical Name or Material N-(2,6-Dimethylphenyl)thiourea
Show More Show Less
Hazard Category H302-H319
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 229-005-8
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.