Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2,6-Dimethylphenylcarbamoylmethyl)iminodiacetic Acid 98.0+%, TCI America™
SDP

Catalog No. D17301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D17301G Supplier TCI America Supplier No. D17301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59160-29-1
Molecular Formula C14H18N2O5
Molecular Weight (g/mol) 294.31
MDL Number MFCD00038399
InChI Key DJQJFMSHHYAZJD-UHFFFAOYSA-N
Synonym N-(2,6-Xylidinocarbonylmethyl)iminodiacetic Acid
PubChem CID 42963
IUPAC Name 2-[(carboxymethyl)({[(2,6-dimethylphenyl)carbamoyl]methyl})amino]acetic acid
SMILES CC1=CC=CC(C)=C1NC(=O)CN(CC(O)=O)CC(O)=O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 294.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2,6-Dimethylphenylcarbamoylmethyl)iminodiacetic Acid
RTECSNumber AH1846000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.