Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(2-Acetamido)-2-aminoethanesulfonic acid | 7365-82-4 | 98.0% | 25 G
SDP

Catalog No. 5000414467
Encompass_Preferred

ACES (N-(2-Acetamido)-2-aminoethanesulfonic acid) is a zwitterionic buffer with a working pH range of 6.8-7.2. It is for research use only and not sold to patients.

  • Molecular weight: 182.20
  • Formula: C₄H₁₀N₂O₄S
  • CAS No.: 7365-82-4
  • Appearance: Solid, white to off-white
  • Shipping at room temperature
  • Storage: Store at room temperature for 3 years. In solvent, store at -80°C for 2 years or -20°C for 1 year.
  • Solubility in H₂O: 41.67 mg/mL (228.70 mM); needs ultrasonic
  • Solubility in PBS: 16.67 mg/mL (91.49 mM); clear solution; needs ultrasonic

Catalog No. 50-004-14467 Supplier Medchemexpress LLC Supplier No. 25GHYD0866
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACES (N-(2-Acetamido)-2-aminoethanesulfonic acid) is a zwitterionic buffer with a working pH range of 6.8-7.2. It is for research use only and not sold to patients.

  • Molecular weight: 182.20
  • Formula: C₄H₁₀N₂O₄S
  • CAS No.: 7365-82-4
  • Appearance: Solid, white to off-white
  • Shipping at room temperature
  • Storage: Store at room temperature for 3 years. In solvent, store at -80°C for 2 years or -20°C for 1 year.
  • Solubility in H₂O: 41.67 mg/mL (228.70 mM); needs ultrasonic
  • Solubility in PBS: 16.67 mg/mL (91.49 mM); clear solution; needs ultrasonic

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.