Learn More
Medchemexpress LLC N-(2-aminoethyl)-2-(naphthalen-1-yl)acetamide | 36321-43-4 | MFCD08529471 | 228.29 | C14H16N2O | 5 G
SDP
Catalog No. 5000282800
Encompass_Preferred
N-(2-aminoethyl)-2-(naphthalen-1-yl)acetamide is supplied as an analytical reference standard intended for qualitative and quantitative research. The compound is reported as an impurity of naphazoline and is used in chromatographic and mass-spectrometry applications such as HPLC, GC, and MS. CAS 36321-43-4.
Catalog No. 50-002-82800 Supplier Medchemexpress LLC Supplier No. HYW1405665G
N-(2-aminoethyl)-2-(naphthalen-1-yl)acetamide is supplied as an analytical reference standard intended for qualitative and quantitative research. The compound is reported as an impurity of naphazoline and is used in chromatographic and mass-spectrometry applications such as HPLC, GC, and MS. CAS 36321-43-4.