Learn More
Medchemexpress LLC N-(2-aminoethyl)-2-(naphthalen-1-yl)acetamide | 36321-43-4 | MFCD08529471 | 95.0% | 228.29 | C14H16N2O | 1 G
SDP
Catalog No. 5000283014
Encompass_Preferred
N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide is a small-molecule analytical standard provided for research and analytical use. Supplier documentation identifies it as an impurity of naphazoline; it is used primarily as a reference material for impurity profiling, method development, and analytical verification in laboratories.
  • Used as an analytical standard for impurity profiling and reference
  • Applicable to qualitative and quantitative methods including HPLC and LC-MS
  • Small, stable solid suitable for laboratory storage and handling
  • Storage: keep at 4°C and protect from light; in solution, follow low-temperature conditions
  • Identified impurity of naphazoline useful for drug-substance analysis
  • Chemical identifiers: CAS 36321-43-4; formula C14H16N2O; molecular weight 228.29
Catalog No. 50-002-83014 Supplier Medchemexpress LLC Supplier No. HYW1405661G
N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide is a small-molecule analytical standard provided for research and analytical use. Supplier documentation identifies it as an impurity of naphazoline; it is used primarily as a reference material for impurity profiling, method development, and analytical verification in laboratories.
  • Used as an analytical standard for impurity profiling and reference
  • Applicable to qualitative and quantitative methods including HPLC and LC-MS
  • Small, stable solid suitable for laboratory storage and handling
  • Storage: keep at 4°C and protect from light; in solution, follow low-temperature conditions
  • Identified impurity of naphazoline useful for drug-substance analysis
  • Chemical identifiers: CAS 36321-43-4; formula C14H16N2O; molecular weight 228.29