Learn More
Medchemexpress LLC N-(2-aminoethyl)-2-(naphthalen-1-yl)acetamide | 36321-43-4 | MFCD08529471 | >=95.0% | 228.29 g·mol⁻¹ | C14H16N2O | 500 MG
SDP
Catalog No. 5000283573
Encompass_Preferred
N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide is an analytical reference standard reported as an impurity of naphazoline. It is intended for use in qualitative and quantitative laboratory analyses and is typically used in method development and validation for chromatographic and mass spectrometric assays. Consult the certificate of analysis and safety data sheet for purity, handling, and storage instructions.
  • Used as an analytical reference standard for HPLC, GC, and MS.
  • Suitable for method development, validation, and impurity profiling.
  • Available in small-scale quantities for laboratory use.
  • Provided with a certificate of analysis detailing purity and storage.
  • Solid form; handle according to safety data sheet recommendations.
Catalog No. 50-002-83573 Supplier Medchemexpress LLC Supplier No. HYW140566500MG
N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide is an analytical reference standard reported as an impurity of naphazoline. It is intended for use in qualitative and quantitative laboratory analyses and is typically used in method development and validation for chromatographic and mass spectrometric assays. Consult the certificate of analysis and safety data sheet for purity, handling, and storage instructions.
  • Used as an analytical reference standard for HPLC, GC, and MS.
  • Suitable for method development, validation, and impurity profiling.
  • Available in small-scale quantities for laboratory use.
  • Provided with a certificate of analysis detailing purity and storage.
  • Solid form; handle according to safety data sheet recommendations.