Learn More
Medchemexpress LLC N-(2-aminoethyl)-2-(naphthalen-1-yl)acetamide | 36321-43-4 | MFCD08529471 | 96.0% | 228.29 g/mol | C14H16N2O | 250 MG
SDP
Catalog No. 5000283561
Encompass_Preferred
N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide is a small-molecule impurity of naphazoline supplied as a research reagent and analytical standard. It is a solid (light brown to brown) with formula C14H16N2O and molecular weight 228.29 g/mol, intended for impurity profiling and analytical method development. Store at 4°C and protect from light; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Used as an analytical reference for impurity identification.
  • Suitable for method development and impurity profiling.
  • Solid form facilitates handling and accurate weighing.
  • Storage recommendations support short- and long-term stability.
  • Supplied in small research quantities for laboratory use.
Catalog No. 50-002-83561 Supplier Medchemexpress LLC Supplier No. HYW140566250MG
N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide is a small-molecule impurity of naphazoline supplied as a research reagent and analytical standard. It is a solid (light brown to brown) with formula C14H16N2O and molecular weight 228.29 g/mol, intended for impurity profiling and analytical method development. Store at 4°C and protect from light; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Used as an analytical reference for impurity identification.
  • Suitable for method development and impurity profiling.
  • Solid form facilitates handling and accurate weighing.
  • Storage recommendations support short- and long-term stability.
  • Supplied in small research quantities for laboratory use.