Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Aminoethyl)-3-aminopropylmethyldimethoxysilane, 95%, Thermo Scientific™

Catalog No. AA4241118
Change view
Click to view available options
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA4241118 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4241118 Supplier Thermo Scientific Chemicals Supplier No. 4241118

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3069-29-2
Quantity 50g
Molecular Formula C8H22N2O2Si
MDL Number MFCD00054776
UN Number UN2735
Flash Point 90°C (194°F)
Synonym n-2-aminoethyl-3-aminopropylmethyldimethoxysilane, 3-2-aminoethylamino propyl-dimethoxymethylsilane, 3-2-aminoethylamino propyldimethoxymethylsilane, 3-2-aminoethylamino propylmethyldimethoxysilane, n-3-dimethoxymethylsilyl propyl ethylenediamine, 1,2-ethanediamine, n-3-dimethoxymethylsilyl propyl, n-3-dimethoxy methyl silyl propyl ethane-1,2-diamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 1,2-ethanediamine, n1-3-dimethoxymethylsilyl propyl
InChI Key MQWFLKHKWJMCEN-UHFFFAOYSA-N
SMILES CO[Si](C)(CCCNCCN)OC
IUPAC Name (2-aminoethyl)({3-[dimethoxy(methyl)silyl]propyl})amine
Molecular Weight (g/mol) 206.36
PubChem CID 76482
Formula Weight 206.36
Percent Purity 95%
Refractive Index 1.4447
Density 0.975g/mL
Chemical Name or Material N- (2-Aminoethyl)-3-aminopropylmethyldimethoxysilane
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.