Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(2-aminoethyl)maleimide hydrochloride | 134272-64-3 | MFCD09800491 | 100.0% | 176.60 g/mol | C6H9ClN2O2 | 5 G
SDP

Catalog No. 5000210758
Encompass_Preferred

N-(2-aminoethyl)maleimide hydrochloride is a thiol-reactive maleimide cross-linker used in bioconjugation and in the preparation of MMP-2-sensitive nanosystems for research applications. It is supplied as the hydrochloride salt and is reported at high purity.

  • Thiol-reactive maleimide cross-linker
  • High purity suitable for research use
  • Molecular weight 176.60 g/mol
  • CAS number 134272-64-3
  • Typically used for bioconjugation and nanosystem synthesis
  • Provided as the hydrochloride salt

Catalog No. 50-002-10758 Supplier Medchemexpress LLC Supplier No. HYW068119A5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-(2-aminoethyl)maleimide hydrochloride is a thiol-reactive maleimide cross-linker used in bioconjugation and in the preparation of MMP-2-sensitive nanosystems for research applications. It is supplied as the hydrochloride salt and is reported at high purity.

  • Thiol-reactive maleimide cross-linker
  • High purity suitable for research use
  • Molecular weight 176.60 g/mol
  • CAS number 134272-64-3
  • Typically used for bioconjugation and nanosystem synthesis
  • Provided as the hydrochloride salt

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.