Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-(2-Aminoethyl)Maleimide Trifluoroacetate Salt

Catalog No. 56951250MGF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-951-250MGF 250 mg
56-951-1GF 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 56-951-250MGF Supplier Sigma Aldrich Supplier No. 56951250MGF
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-(2-Aminoethyl)maleimide trifluoroacetate salt may can be to synthesize:N-(2-{3-iodobenzoyl}aminoethyl)maleimideN-(2-{3-(tri-n-butylstannyl)benzoyl}aminoethyl)maleimidemaleimide-functionalized cymantrenyl derivativesmaleimide-incorporated anisotropic biohybrid microparticles (maleimide-MPs)Heterobifunctional linker for introducing a maleimide function into the widely used NOSu-modified fluorescent labels.

Specifications

CAS 146474-00-2
Molecular Formula C6H8N2O2 -+ C2HF3O2
Linear Formula C6H8N2O2 -+ C2HF3O2
MDL Number MFCD01073617
Quantity 250 mg
Synonym 2-Maleimidoethylamine trifluoroacetate salt
Molecular Weight (g/mol) 254.16 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.