Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(2-aminoethyl)piperazine-1,4-diethylamine | 31295-54-2 | MFCD01941014 | ≥97.0% | 215.34 g/mol | C10H25N5 | 250 MG
SDP

Catalog No. 5000281816
Encompass_Preferred

N-(2-Aminoethyl)piperazine-1,4-diethylamine is a small-molecule biochemical reagent supplied for laboratory research use. It is used as an organic building block or reagent in medicinal chemistry and biochemical studies. For research use only; not for clinical, diagnostic, or therapeutic applications.

  • Small-molecule biochemical reagent for research applications.
  • Purity ≥97.0%.
  • Molecular formula C10H25N5; molecular weight 215.34 g/mol.
  • Appearance: colorless to light yellow liquid.
  • Storage: 4°C, protect from light; in solvent, -80°C (6 months) or -20°C (1 month).
  • CAS number 31295-54-2.

Catalog No. 50-002-81816 Supplier Medchemexpress LLC Supplier No. HYW265730250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-(2-Aminoethyl)piperazine-1,4-diethylamine is a small-molecule biochemical reagent supplied for laboratory research use. It is used as an organic building block or reagent in medicinal chemistry and biochemical studies. For research use only; not for clinical, diagnostic, or therapeutic applications.

  • Small-molecule biochemical reagent for research applications.
  • Purity ≥97.0%.
  • Molecular formula C10H25N5; molecular weight 215.34 g/mol.
  • Appearance: colorless to light yellow liquid.
  • Storage: 4°C, protect from light; in solvent, -80°C (6 months) or -20°C (1 month).
  • CAS number 31295-54-2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.