Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(2-aminoethyl)piperazine-1,4-diethylamine | 31295-54-2 | MFCD01941014 | ≥97.0% | 215.34 g/mol | C10H25N5 | 500 MG
SDP

Catalog No. 5000282088
Encompass_Preferred

N-(2-aminoethyl)piperazine-1,4-diethylamine is a piperazine-derived diamine used as a biochemical reagent and synthetic intermediate in life-science research. It is supplied as a liquid and characterized by analytical data that confirm identity and suitability for research applications.

  • CAS number: 31295-54-2.
  • Purity: ≥97.0%.
  • Molecular formula: C10H25N5.
  • Molecular weight: 215.34 g/mol.
  • Appearance: colorless to light yellow liquid.
  • Analytical data: 1H NMR and LCMS consistent with structure.
  • Typical pack size: 500 MG.

Catalog No. 50-002-82088 Supplier Medchemexpress LLC Supplier No. HYW265730500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-(2-aminoethyl)piperazine-1,4-diethylamine is a piperazine-derived diamine used as a biochemical reagent and synthetic intermediate in life-science research. It is supplied as a liquid and characterized by analytical data that confirm identity and suitability for research applications.

  • CAS number: 31295-54-2.
  • Purity: ≥97.0%.
  • Molecular formula: C10H25N5.
  • Molecular weight: 215.34 g/mol.
  • Appearance: colorless to light yellow liquid.
  • Analytical data: 1H NMR and LCMS consistent with structure.
  • Typical pack size: 500 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.