Learn More
AdipoGen N-2-Aminoethylbiotinamid HCl
SDP
Catalog No. 502561804
Encompass_Preferred
Chemical. CAS 111822-45-8. Formula C12H23ClN4O2S. MW 322.85. Synthetic. Amino derivative of biotin that can be used as an intracellular label for cells, particularly neurons. It is used for neuronal tracing studies by visualizing neural architecture and for the identification of gap junction coupling. It is better soluble and non-toxic compared to other neuronal labels. It remains longer in cellsand can be fixed with formalin or glutaraldehyde. It can be detected using avidin or streptavidin systems with either chromogenic or fluorescence visualization methods.
Catalog No. 50-256-1804 Supplier AdipoGen Supplier No. CDXA0191M050
Chemical. CAS 111822-45-8. Formula C12H23ClN4O2S. MW 322.85. Synthetic. Amino derivative of biotin that can be used as an intracellular label for cells, particularly neurons. It is used for neuronal tracing studies by visualizing neural architecture and for the identification of gap junction coupling. It is better soluble and non-toxic compared to other neuronal labels. It remains longer in cellsand can be fixed with formalin or glutaraldehyde. It can be detected using avidin or streptavidin systems with either chromogenic or fluorescence visualization methods.