Learn More
Abcam N-(2-Aminoethyl)biotinamide hydrochloride, biotin for intracellular labeling, 50MG
Catalog No. 502705202
Encompass_Preferred
MW 322.85 g/mol, Purity >95%. Amino derivative of biotin that can be used as an intracellular label for cells, particularly neurons. Used for neuronal tracing studies by visualizing neural architecture and for the identification of gap junction coupling. More soluble and non-toxic compared to other neuronal labels. Remains longer in cells and can be fixed with formalin or glutaraldehyde. Can be detected using avidin or streptavidin systems with either chromogenic or fluorescence visualization methods.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5202 Supplier Abcam Supplier No. AB14542350MG
MW 322.85 g/mol, Purity >95%. Amino derivative of biotin that can be used as an intracellular label for cells, particularly neurons. Used for neuronal tracing studies by visualizing neural architecture and for the identification of gap junction coupling. More soluble and non-toxic compared to other neuronal labels. Remains longer in cells and can be fixed with formalin or glutaraldehyde. Can be detected using avidin or streptavidin systems with either chromogenic or fluorescence visualization methods.
The product is subject to the following: Abcam Restricted Use Statement