Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Carboxybenzoyl)-(-)-10,2-camphorsultam 98.0+%, TCI America™
SDP

Catalog No. C1682500MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1682500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C1682500MG Supplier TCI America Supplier No. C1682500MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 179950-32-4
Molecular Formula C18H21NO5S
Molecular Weight (g/mol) 363.43
MDL Number MFCD04117913
InChI Key UJCSAJNSMWSFHF-UHFFFAOYNA-N

Specifications

Melting Point 183°C
Color White
Quantity 500 mg
Formula Weight 363.43
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Carboxybenzoyl)-(-)-10,2-camphorsultam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.