Learn More
Medchemexpress LLC N-(2-carboxyphenyl)-4-chloroanthranilic acid disodium salt | 64808-48-6 | MFCD00941422 | 99.9% | 335.65 | C14H8ClNNa2O4 | 100 MG
SDP
Catalog No. 5000241309
Encompass_Preferred
Lobenzarit disodium is the disodium salt form of lobenzarit, an immunomodulatory compound with anti-arthritic and antioxidative activity. Supplied as a high-purity solid for research use, it is used in studies of immune modulation, inflammation, and oxidative stress, and is suitable for formulation in aqueous media for in vitro and exploratory in vivo work.
  • High purity suitable for research applications.
  • Molecular weight 335.65 for accurate dosing calculations.
  • Disodium salt form improves water solubility and handling.
  • Available in multiple small package sizes for dose flexibility.
  • Commonly used in immunomodulation and anti-arthritic studies.
  • Provided with batch-specific certificate of analysis.
Catalog No. 50-002-41309 Supplier Medchemexpress LLC Supplier No. HY107360100MG
Lobenzarit disodium is the disodium salt form of lobenzarit, an immunomodulatory compound with anti-arthritic and antioxidative activity. Supplied as a high-purity solid for research use, it is used in studies of immune modulation, inflammation, and oxidative stress, and is suitable for formulation in aqueous media for in vitro and exploratory in vivo work.
  • High purity suitable for research applications.
  • Molecular weight 335.65 for accurate dosing calculations.
  • Disodium salt form improves water solubility and handling.
  • Available in multiple small package sizes for dose flexibility.
  • Commonly used in immunomodulation and anti-arthritic studies.
  • Provided with batch-specific certificate of analysis.